CH4424851625 ChemBridge Corporation

(1,3-benzodioxol-5-ylmethyl)(2-pyridinylmethyl)amine dihydrobromide

  • MDL number MFCD26959642
  • Cas Number : 1609406-32-7
  • Molecular Weight : 404.097

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5552710

Safety Information

MSDS https://www.hit2lead.com/sds/5552710_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart