CH4424851629 ChemBridge Corporation

[2-(1H-indol-3-yl)ethyl](3-pyridinylmethyl)amine hydrobromide

  • MDL number MFCD13194069
  • Cas Number : 1609409-22-4
  • Molecular Weight : 332.2382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5553645

Safety Information

MSDS https://www.hit2lead.com/sds/5553645_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart