CH4424851637 ChemBridge Corporation

(4-methoxybenzyl)(2-methylbenzyl)amine hydrobromide

  • MDL number MFCD13186453
  • Cas Number : 1609395-74-5
  • Molecular Weight : 322.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5554908

Safety Information

MSDS https://www.hit2lead.com/sds/5554908_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart