CH4424851639 ChemBridge Corporation

N-(2-methoxybenzyl)-2-methylcyclohexanamine hydrobromide

  • MDL number MFCD13186455
  • Cas Number : 1609407-26-2
  • Molecular Weight : 314.2612

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5555095

Safety Information

MSDS https://www.hit2lead.com/sds/5555095_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart