CH4424851641 ChemBridge Corporation

N-(2-pyridinylmethyl)-1-adamantanamine hydrobromide

  • MDL number MFCD09870088
  • Cas Number : 1609408-97-0
  • Molecular Weight : 323.2712

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5555333

Safety Information

MSDS https://www.hit2lead.com/sds/5555333_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart