CH4424851642 ChemBridge Corporation

N-(3-methoxybenzyl)-1-(2-methoxyphenyl)methanamine hydrobromide

  • MDL number MFCD26959385
  • Cas Number : 1609399-91-8
  • Molecular Weight : 338.2395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5555346

Safety Information

MSDS https://www.hit2lead.com/sds/5555346_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart