CH4424851646 ChemBridge Corporation

N-(4-methoxybenzyl)-1-(2-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD06800900
  • Cas Number : 436099-93-3
  • Molecular Weight : 293.7885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5556371

Safety Information

MSDS https://www.hit2lead.com/sds/5556371_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart