CH4424851649 ChemBridge Corporation

N-(2,4,5-trimethoxybenzyl)cyclopentanamine hydrobromide

  • MDL number MFCD13186264
  • Cas Number : 1609395-84-7
  • Molecular Weight : 346.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5556822

Safety Information

MSDS https://www.hit2lead.com/sds/5556822_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart