CH4424851650 ChemBridge Corporation

N-(4-nitrobenzyl)cyclohexanamine hydrobromide

  • MDL number MFCD13186265
  • Cas Number : 1609409-25-7
  • Molecular Weight : 315.2062

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5556831

Safety Information

MSDS https://www.hit2lead.com/sds/5556831_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart