CH4424851656 ChemBridge Corporation

[2-(4-methoxyphenyl)ethyl](2-pyridinylmethyl)amine hydrobromide

  • MDL number MFCD13193976
  • Cas Number : 1609409-48-4
  • Molecular Weight : 323.2282

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5558316

Safety Information

MSDS https://www.hit2lead.com/sds/5558316_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart