CH4424851661 ChemBridge Corporation

(2-methoxy-1-methylethyl)(3-methylbenzyl)amine hydrobromide

  • MDL number MFCD06800928
  • Cas Number : 1052529-83-5
  • Molecular Weight : 274.1973

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5559382

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5559382
Availability details for
Quantity
  • Availability
  • Add to Cart