CH4424851663 ChemBridge Corporation

[(7-hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-2-yl)thio]acetic acid hydrate

  • MDL number MFCD26959605
  • Cas Number : 1609406-34-9
  • Molecular Weight : 258.2544

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5560222

Safety Information

MSDS https://www.hit2lead.com/sds/5560222_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart