CH4424851667 ChemBridge Corporation

(5-bromo-2-methoxybenzyl)(4-methoxybenzyl)amine hydrobromide

  • MDL number MFCD13186490
  • Cas Number : 1609404-35-4
  • Molecular Weight : 417.1356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561082

Safety Information

MSDS https://www.hit2lead.com/sds/5561082_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart