CH4424851668 ChemBridge Corporation

N-(2,5-dimethoxybenzyl)-2-phenylethanamine hydrobromide

  • MDL number MFCD13186491
  • Cas Number : 1609407-57-9
  • Molecular Weight : 352.2661

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561090

Safety Information

MSDS https://www.hit2lead.com/sds/5561090_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart