CH4424851672 ChemBridge Corporation

N-(3,5-dimethoxybenzyl)-2-propanamine hydrobromide

  • MDL number MFCD13186494
  • Cas Number : 1609406-46-3
  • Molecular Weight : 290.1967

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561898

Safety Information

MSDS https://www.hit2lead.com/sds/5561898_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart