CH4424851673 ChemBridge Corporation

(4-methoxybenzyl)(2-pyridinylmethyl)amine hydrobromide

  • MDL number MFCD13194075
  • Cas Number : 1609400-07-8
  • Molecular Weight : 309.2016

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561926

Safety Information

MSDS https://www.hit2lead.com/sds/5561926_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart