CH4424851674 ChemBridge Corporation

[2-(4-fluorophenyl)ethyl](4-nitrobenzyl)amine hydrobromide

  • MDL number MFCD13186495
  • Cas Number : 1609400-18-1
  • Molecular Weight : 355.2021

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561968

Safety Information

MSDS https://www.hit2lead.com/sds/5561968_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart