CH4424851678 ChemBridge Corporation

(4-fluorobenzyl)(2-methoxybenzyl)amine hydrobromide

  • MDL number MFCD13186498
  • Cas Number : 1609406-51-0
  • Molecular Weight : 326.204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5562589

Safety Information

MSDS https://www.hit2lead.com/sds/5562589_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart