CH4424851680 ChemBridge Corporation

(2-methoxy-1-methylethyl)(1-naphthylmethyl)amine hydrobromide

  • MDL number MFCD13186499
  • Cas Number : 1609407-58-0
  • Molecular Weight : 310.2294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5563804

Safety Information

MSDS https://www.hit2lead.com/sds/5563804_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart