CH4424851685 ChemBridge Corporation

[2-(3,4-dimethoxyphenyl)ethyl](3-fluorobenzyl)amine hydrobromide

  • MDL number MFCD13186503
  • Cas Number : 1609395-98-3
  • Molecular Weight : 370.2565

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5564882

Safety Information

MSDS https://www.hit2lead.com/sds/5564882_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart