CH4424851687 ChemBridge Corporation

N-(3,5-dimethoxybenzyl)-1-methoxy-2-propanamine hydrobromide

  • MDL number MFCD13186505
  • Cas Number : 1609403-45-3
  • Molecular Weight : 320.2227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/5565122

Safety Information

msds https://www.hit2lead.com/building-blocks/5565122
Availability details for
Quantity
  • Availability
  • Add to Cart