CH4424851691 ChemBridge Corporation

(2-methoxybenzyl)(3-methylbenzyl)amine hydrobromide

  • MDL number MFCD13186508
  • Cas Number : 1609406-41-8
  • Molecular Weight : 322.2401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/5566088

Safety Information

msds https://www.hit2lead.com/building-blocks/5566088
Availability details for
Quantity
  • Availability
  • Add to Cart