CH4424851694 ChemBridge Corporation

N-(2-methoxybenzyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186511
  • Cas Number : 1609407-77-3
  • Molecular Weight : 288.2239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5566942

Safety Information

MSDS https://www.hit2lead.com/sds/5566942_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart