CH4424851703 ChemBridge Corporation

N-(2-methoxybenzyl)-1-(4-methoxy-2,5-dimethylphenyl)methanamine hydrobromide

  • MDL number MFCD26959375
  • Cas Number : 1609403-97-5
  • Molecular Weight : 366.2927

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5568465

Safety Information

MSDS https://www.hit2lead.com/sds/5568465_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart