CH4424851704 ChemBridge Corporation

N-(2,3,4-trimethoxybenzyl)cyclopentanamine hydrobromide

  • MDL number MFCD13186518
  • Cas Number : 1609408-86-7
  • Molecular Weight : 346.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5568829

Safety Information

MSDS https://www.hit2lead.com/sds/5568829_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart