CH4424851707 ChemBridge Corporation

N-(4-methoxybenzyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrobromide

  • MDL number MFCD13186473
  • Cas Number : 1609399-95-2
  • Molecular Weight : 348.2774

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5569275

Safety Information

MSDS https://www.hit2lead.com/sds/5569275_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart