CH4424851708 ChemBridge Corporation

N-[(4-methoxy-1-naphthyl)methyl]-2-propanamine hydrobromide

  • MDL number MFCD13186474
  • Cas Number : 1983847-95-5
  • Molecular Weight : 310.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5569371

Safety Information

MSDS https://www.hit2lead.com/sds/5569371_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart