CH4424851711 ChemBridge Corporation

N-(4-nitrobenzyl)cyclopentanamine hydrochloride

  • MDL number MFCD07109735
  • Cas Number : 1158425-54-7
  • Molecular Weight : 256.7286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5570287

Safety Information

MSDS https://www.hit2lead.com/sds/5570287_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart