CH4424851712 ChemBridge Corporation

(3,4-dimethoxybenzyl)(2-fluorobenzyl)amine hydrobromide

  • MDL number MFCD13186476
  • Cas Number : 1609407-61-5
  • Molecular Weight : 356.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5570292

Safety Information

MSDS https://www.hit2lead.com/sds/5570292_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart