CH4424851727 ChemBridge Corporation

N-[2-(3,4-dimethoxyphenyl)ethyl]cyclohexanamine hydrobromide

  • MDL number MFCD13186526
  • Cas Number : 1609407-37-5
  • Molecular Weight : 344.2871

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5573007

Safety Information

MSDS https://www.hit2lead.com/sds/5573007_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart