CH4424851734 ChemBridge Corporation

(2-methoxybenzyl)(3-nitrobenzyl)amine hydrobromide

  • MDL number MFCD13186532
  • Cas Number : 1609400-22-7
  • Molecular Weight : 353.2111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5574151

Safety Information

MSDS https://www.hit2lead.com/sds/5574151_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart