CH4424851749 ChemBridge Corporation

N-(3-nitrobenzyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrobromide

  • MDL number MFCD13186468
  • Cas Number : 1609400-35-2
  • Molecular Weight : 363.249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5576492

Safety Information

MSDS https://www.hit2lead.com/sds/5576492_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart