CH4424851768 ChemBridge Corporation

(2-methoxyethyl)[2-(2-naphthyloxy)ethyl]amine hydrochloride

  • MDL number MFCD13186537
  • Cas Number : 1609408-89-0
  • Molecular Weight : 281.7778

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5733605

Safety Information

MSDS https://www.hit2lead.com/sds/5733605_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart