CH4424851769 ChemBridge Corporation

2-{[2-(4-chlorophenoxy)ethyl]amino}ethanol hydrochloride

  • MDL number MFCD13186538
  • Cas Number : 1609400-24-9
  • Molecular Weight : 252.1376

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5737767

Safety Information

MSDS https://www.hit2lead.com/sds/5737767_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart