CH4424851776 ChemBridge Corporation

(3-ethyl-2-imino-2,3-dihydro-1H-benzimidazol-1-yl)acetic acid hydrobromide

  • MDL number MFCD02106443
  • Cas Number : 436099-71-7
  • Molecular Weight : 300.1518

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5785978

Safety Information

MSDS https://www.hit2lead.com/sds/5785978_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart