CH4424851777 ChemBridge Corporation

1-(1-adamantyl)-2-(isopropylamino)ethanone hydrochloride

  • MDL number MFCD01909245
  • Cas Number : 33447-62-0
  • Molecular Weight : 271.826

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5807100

Safety Information

MSDS https://www.hit2lead.com/sds/5807100_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart