CH4424851780 ChemBridge Corporation

3-[(2-aminoethyl)thio]-1-(4-methoxyphenyl)-2,5-pyrrolidinedione hydrochloride

  • MDL number MFCD01975222
  • Cas Number : 1047652-78-7
  • Molecular Weight : 316.8037

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5851732

Safety Information

MSDS https://www.hit2lead.com/sds/5851732_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart