CH4424851781 ChemBridge Corporation

3-[(2-aminoethyl)thio]-1-(4-fluorophenyl)-2,5-pyrrolidinedione hydrochloride

  • MDL number MFCD03941512
  • Cas Number : 1048649-31-5
  • Molecular Weight : 304.7682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5854724

Safety Information

MSDS https://www.hit2lead.com/sds/5854724_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart