CH4424851784 ChemBridge Corporation

N-benzyl-6-methyl-2,3,4,9-tetrahydro-1H-carbazol-1-amine hydrochloride

  • MDL number MFCD00563898
  • Cas Number : 115174-34-0
  • Molecular Weight : 326.863

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5869604

Safety Information

MSDS https://www.hit2lead.com/sds/5869604_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart