CH4424851786 ChemBridge Corporation

9-ethyl-3-(1-piperazinylmethyl)-9H-carbazole dihydrochloride

  • MDL number MFCD13186541
  • Cas Number : 1609407-79-5
  • Molecular Weight : 366.3279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5878848

Safety Information

MSDS https://www.hit2lead.com/sds/5878848_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart