CH4424851787 ChemBridge Corporation

1-(2-bromobenzyl)piperazine hydrochloride

  • MDL number MFCD13186542
  • Cas Number : 1063613-12-6
  • Molecular Weight : 291.6151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5880609

Safety Information

MSDS https://www.hit2lead.com/sds/5880609_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart