CH4424851794 ChemBridge Corporation

N-(3-chlorobenzyl)-2-(1H-indol-3-yl)ethanamine hydrobromide

  • MDL number MFCD13186482
  • Cas Number : 1609400-49-8
  • Molecular Weight : 365.6952

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5937338

Safety Information

MSDS https://www.hit2lead.com/sds/5937338_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart