CH4424851797 ChemBridge Corporation

N-(4-ethylbenzyl)-2-(4-methoxyphenyl)ethanamine hydrobromide

  • MDL number MFCD13186547
  • Cas Number : 1609401-37-7
  • Molecular Weight : 350.2933

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5937667

Safety Information

MSDS https://www.hit2lead.com/sds/5937667_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart