CH4424851800 ChemBridge Corporation

(1,3-benzodioxol-5-ylmethyl)(2-chlorobenzyl)amine hydrobromide

  • MDL number MFCD13186549
  • Cas Number : 1609404-15-0
  • Molecular Weight : 356.6421

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5938632

Safety Information

MSDS https://www.hit2lead.com/sds/5938632_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart