CH4424851801 ChemBridge Corporation

1-benzyl-N-(1-ethylpropyl)-4-piperidinamine dihydrobromide

  • MDL number MFCD13186550
  • Cas Number : 1609409-50-8
  • Molecular Weight : 422.2415

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5938636

Safety Information

MSDS https://www.hit2lead.com/sds/5938636_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart