CH4424851804 ChemBridge Corporation

N-(sec-butyl)cyclopentanamine hydrobromide

  • MDL number MFCD09870091
  • Cas Number : 1609408-90-3
  • Molecular Weight : 222.1658

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5938899

Safety Information

MSDS https://www.hit2lead.com/sds/5938899_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart