CH4424851818 ChemBridge Corporation

(2-chlorobenzyl)(3,4-dimethoxybenzyl)amine hydrobromide

  • MDL number MFCD13186568
  • Cas Number : 1609403-48-6
  • Molecular Weight : 372.6846

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5943316

Safety Information

MSDS https://www.hit2lead.com/sds/5943316_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart