CH4424851820 ChemBridge Corporation

N-(2-chlorobenzyl)-2-(4-methoxyphenyl)ethanamine hydrobromide

  • MDL number MFCD13186570
  • Cas Number : 1609403-96-4
  • Molecular Weight : 356.6852

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5944887

Safety Information

MSDS https://www.hit2lead.com/sds/5944887_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart