CH4424851830 ChemBridge Corporation

(3-chlorobenzyl)(3,4-dimethoxybenzyl)amine hydrobromide

  • MDL number MFCD13186565
  • Cas Number : 1609400-86-3
  • Molecular Weight : 372.6846

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5947140

Safety Information

MSDS https://www.hit2lead.com/sds/5947140_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart