CH4424851831 ChemBridge Corporation

N-(2-phenylethyl)cyclohexanamine hydrochloride

  • MDL number MFCD00156209
  • Cas Number : 197357-53-2
  • Molecular Weight : 239.7842

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5947232

Safety Information

MSDS https://www.hit2lead.com/sds/5947232_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart